Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000246895.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000381060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000506576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000507962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000510010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000511658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
ENST00000615994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_001009181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007770.hg.1 |
NM_003154.2 |
Refseq |