Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8017345 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8017344 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000484823.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000484823.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000484823.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000484823.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011371.hg.1 |
ENST00000624147.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
241180_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
241180_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
241180_at8 |
- |
- |
Human Genome U133 Set |
241180_at |
- |
- |
Human Genome U133 Set |
241180_PM_at |
- |
- |