Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131221.1 | - | 1 | 1849 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleosome assembly protein 1 like 4 | NAP1L4 |
NAP1L4b, NAP2, NAP2L, hNAP2 |
-63.0kb | Sense | Chr. 11 - 2944430 - 2992377 on NCBI Build GRCh38 |
Downstream | oxysterol binding protein like 5 | OSBPL5 |
OBPH1, ORP5 |
29.7kb | Sense | Chr. 11 - 3087116 - 3165352 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |