Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRO0471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF111846.2 | - | 1 | 1487 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MBNL1 antisense RNA 1 | MBNL1-AS1 | -60.2kb | Anti-sense | Chr. 3 - 152262616 - 152269626 on NCBI Build GRCh38 | |
Downstream |
transmembrane protein 14E, pseudogene |
TMEM14EP | TMEM14E | 8.1kb | Anti-sense | Chr. 3 - 152339698 - 152340990 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |