Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC119664.1 | - | 2 - 3 | 700 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | A kinase (PRKA) anchor inhibitor 1 | AKAIN1 | C18orf42 | -37.6kb | Anti-sense | Chr. 18 - 5143673 - 5199997 on NCBI Build GRCh38 |
Downstream | zinc finger and BTB domain containing 14 | ZBTB14 |
ZF5, ZFP-161, ZFP-5, ZFP161, ZNF478 |
50.5kb | Anti-sense | Chr. 18 - 5289019 - 5297053 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | NR_026849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | NR_026849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | NR_026849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | NR_026849.1 | Refseq |