Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF161370.1 | - | 1 | 244 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928618 | LOC101928618 | -180.4kb | Sense | Chr. 7 - 36095310 - 36100653 on NCBI Build GRCh38 | |
Downstream | KIAA0895 | KIAA0895 | 41.7kb | Anti-sense | Chr. 7 - 36324150 - 36390125 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |