Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128687.1 | - | 1 | 3153 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |