Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB2B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC114619.1 | - | 1 | 1244 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
secretoglobin family 1B member 2, pseudogene |
SCGB1B2P | SCGB4A1P | -67.7kb | Anti-sense | Chr. 19 - 34576733 - 34677031 on NCBI Build GRCh38 |
Downstream | zinc finger protein 302 | ZNF302 |
HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327 |
30.8kb | Sense | Chr. 19 - 34677619 - 34686397 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |