Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100159195.hg.1 |
TC2100008576.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100159195.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100159195.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100159195.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100159195.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
ENST00000400372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008576.hg.1 |
NM_198694.3 |
Refseq |