Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002016.hg.1 |
ENST00000399387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002016.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002016.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002016.hg.1 |
TC14000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
ENST00000399387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
ENST00000548213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
ENST00000552957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002018.hg.1 |
TC14000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
ENST00000399387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
ENST00000548213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
ENST00000552957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002019.hg.1 |
TC14000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002024.hg.1 |
ENST00000399387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002024.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002024.hg.1 |
TC14000185.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147185.hg.1 |
TC1400006792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000550266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006792.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000548213.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
ENST00000552957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147186.hg.1 |
TC1400006792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147191.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147191.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147191.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147191.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147191.hg.1 |
TC1400006792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
ENST00000399387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
NR_026731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
NR_026732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147182.hg.1 |
TC1400006792.hg.1 |
Clariom D TC |