Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007524.hg.1 |
TC22001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007524.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007524.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007520.hg.1 |
TC22001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007520.hg.1 |
ENST00000381051.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007520.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
TC2200009286.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169367.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009286.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
TC2200009286.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
NR_027034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000416202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000445441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
ENST00000451166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169372.hg.1 |
NR_027034.1 |
Refseq |