Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031374.hg.1 |
TC01002075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031374.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031374.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031374.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000427528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009311.hg.1 |
ENST00000522984.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009311.hg.1 |
ENST00000522984.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031375.hg.1 |
TC01002075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031375.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031375.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031375.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031375.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009504.hg.1 |
ENST00000411632.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009504.hg.1 |
ENST00000411632.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000423888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000427528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000462238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000463325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000467796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000480246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000494075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
ENST00000496652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007828.hg.1 |
NR_026982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191192.hg.1 |
TC0100012421.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
ENST00000427857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
ENST00000446136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
NR_027055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100191191.hg.1 |
TC0100012421.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009311.hg.1 |
ENST00000522984.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009311.hg.1 |
ENST00000522984.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009504.hg.1 |
ENST00000411632.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009504.hg.1 |
ENST00000411632.2 |
Ensembl |