Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
TC09001556.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
ENST00000417201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019559.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011432.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000417201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000483428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
ENST00000491850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NM_033117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NR_027125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
NR_027126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177963.hg.1 |
TC0900011432.hg.1 |
Clariom D TC |