Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012181.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012181.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012181.hg.1 |
TC17000926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009157.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000414089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
ENST00000571031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
NR_027255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700169414.hg.1 |
TC1700009157.hg.1 |
Clariom D TC |