Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7904466 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7919611 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7904468 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7919588 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7904467 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7919587 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7919610 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7919606 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7904465 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7919584 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
ENST00000412169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015506.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016567.hg.1 |
TC01001046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016567.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016567.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016566.hg.1 |
TC01001046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016566.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016566.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
ENST00000412169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015506.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215809.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215809.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215809.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215809.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215809.hg.1 |
TC0100015506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
ENST00000412169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
ENST00000412169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
ENST00000430394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
NR_027337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215811.hg.1 |
TC0100015506.hg.1 |
Clariom D TC |