Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027391.1 | 4 - 5 | 1343 | 92 | ||
GenBank mRNA | BC025710.1 | - | 4 - 5 | 1203 | 5224878 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |