Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000465978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
ENST00000465978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002743.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002743.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002743.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002743.hg.1 |
TC21000229.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002742.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002742.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002742.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002742.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002742.hg.1 |
TC21000229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000465978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
ENST00000465978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007371.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
TC2100007371.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
ENST00000330490.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149892.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
NR_026548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
TC2100007371.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
ENST00000354333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
NR_026547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149893.hg.1 |
NR_026548.1 |
Refseq |