Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159352.1 | NP_001152824.1 | 5 - 6 | 745 | 85 | |
NM_032044.3 | NP_114433.1 | 4 - 5 | 571 | 85 | ||
GenBank mRNA | AF254415.1 | - | 4 - 5 | 486 | 85 | |
AF345934.1 | - | 4 - 5 | 439 | 85 | ||
AY007243.1 | - | 4 - 5 | 427 | 85 | ||
AY126670.1 | - | 5 - 6 | 745 | 85 | ||
BC017089.2 | - | 4 - 5 | 443 | 3850358 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |