Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164407.1 | NP_001157879.1 | 4 | 3573 | 86 | |
GenBank mRNA | AK000038.1 | - | 1 | 546 | 86 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pre-mRNA processing factor 8 | PRPF8 |
HPRP8, PRP8, PRPC8, RP13, SNRNP220 |
-20.2kb | Sense | Chr. 17 - 1650629 - 1684882 on NCBI Build GRCh38 |
Downstream | MIR22 host gene | MIR22HG | C17orf91 | 5.8kb | Sense | Chr. 17 - 1711504 - 1716272 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |