Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
CTAGE family member 3, pseudogene |
CTAGE3P |
CTAGE-3, CTAGE3 |
-71.5kb | Sense | Chr. 13 - 51907820 - 51910559 on NCBI Build GRCh38 |
Downstream |
ALG11, alpha-1,2-mannosyltransferase |
ALG11 |
CDG1P, GT8 |
28.5kb | Anti-sense | Chr. 13 - 52012387 - 52029648 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |