Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PRH1-PRR4 readthrough | PRH1-PRR4 | PRH1 | Sense | Chr. 12 - 10845849 - 11171625 on NCBI Build GRCh38 | |
Downstream | taste 2 receptor member 19 | TAS2R19 |
MSTP058, T2R19, T2R23, T2R48, TAS2R23, TAS2R48 |
13.7kb | Sense | Chr. 12 - 11021619 - 11022620 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |