Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144970.2 | NP_659407.1 | 7 | 4047 | 110 | |
XM_005272589.2 | XP_005272646.2 | 7 | 2461 | 110 | ||
XM_006724527.3 | XP_006724590.2 | 6 | 3867 | 110 | ||
XM_006724528.2 | XP_006724591.1 | 6 | 3892 | 110 | ||
XM_017029302.1 | XP_016884791.1 | 6 | 4038 | 110 | ||
XM_017029303.1 | XP_016884792.1 | 6 | 2306 | 110 | ||
XM_017029304.1 | XP_016884793.1 | 7 | 5209 | 110 | ||
XM_017029305.1 | XP_016884794.1 | 7 | 2496 | 110 | ||
GenBank mRNA | AK021629.1 | - | 1 | 1149 | 111 | |
BM969708.1 | - | 1 | 110 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial pyruvate carrier 1-like | MPC1L | -2.8kb | Anti-sense | Chr. X - 40623566 - 40624139 on NCBI Build GRCh38 | |
Downstream | mediator complex subunit 14 | MED14 |
CRSP150, CRSP2, CSRP, CXorf4, DRIP150, EXLM1, RGR1, TRAP170 |
21.3kb | Sense | Chr. X - 40649543 - 40736122 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |