Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PH domain and leucine rich repeat protein phosphatase 1 | PHLPP1 |
PHLPP, PLEKHE1, PPM3A, SCOP |
-333.4kb | Anti-sense | Chr. 18 - 62715439 - 62980443 on NCBI Build GRCh38 |
Downstream | 3-ketodihydrosphingosine reductase | KDSR |
DHSR, FVT1, SDR35C1 |
13.7kb | Sense | Chr. 18 - 63327736 - 63367273 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |