Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
golgin, RAB6 interacting |
GORAB |
GO, NTKLBP1, SCYL1BP1 |
-152.9kb | Sense | Chr. 1 - 170532122 - 170553834 on NCBI Build GRCh38 |
Downstream | maestro heat like repeat family member 9 | MROH9 |
ARMC11, C1orf129 |
228.1kb | Sense | Chr. 1 - 170935471 - 171066341 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |