Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ008150.1 | - | 1 | 56 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAP3K14 antisense RNA 1 | MAP3K14-AS1 | -2.5kb | Anti-sense | Chr. 17 - 45247925 - 45268630 on NCBI Build GRCh38 | |
Downstream | Rho GTPase activating protein 27 | ARHGAP27 |
CAMGAP1, PP905, SH3D20, SH3P20 |
122.6kb | Sense | Chr. 17 - 45393902 - 45432916 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |