Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GRB10 interacting GYF protein 1 | GIGYF1 |
GYF1, PERQ1 |
Anti-sense | Chr. 7 - 100679507 - 100694282 on NCBI Build GRCh38 | |
Downstream |
POP7 homolog, ribonuclease P/MRP subunit |
POP7 |
0610037N12Rik, RPP2, RPP20 |
15.7kb | Sense | Chr. 7 - 100706053 - 100707500 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |