Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF507951.1 | - | 1 | 848 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TEK receptor tyrosine kinase | TEK |
CD202B, TIE-2, TIE2, VMCM, VMCM1 |
-48.0kb | Anti-sense | Chr. 9 - 27109141 - 27230178 on NCBI Build GRCh38 |
Downstream | equatorin | EQTN |
AFAF, C9orf11, SPACA8 |
4.9kb | Sense | Chr. 9 - 27284654 - 27297139 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |