Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
transmembrane protein 14E, pseudogene |
TMEM14EP | TMEM14E | -112.4kb | Anti-sense | Chr. 3 - 152339698 - 152340990 on NCBI Build GRCh38 |
Downstream | purinergic receptor P2Y1 | P2RY1 | P2Y1 | 379.4kb | Sense | Chr. 3 - 152834693 - 152841439 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |