Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 16 | METTL16 | METT10D | Sense | Chr. 17 - 2416049 - 2511906 on NCBI Build GRCh38 | |
Downstream | platelet activating factor acetylhydrolase 1b regulatory subunit 1 | PAFAH1B1 |
LIS1, LIS2, MDCR, MDS, NudF, PAFAH |
160.0kb | Anti-sense | Chr. 17 - 2593210 - 2685617 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |