Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dopamine receptor D2 | DRD2 |
D2DR, D2R |
-195.3kb | Sense | Chr. 11 - 113409595 - 113475279 on NCBI Build GRCh38 |
Downstream |
transmembrane protease, serine 5 |
TMPRSS5 | SPINESIN | 14.3kb | Sense | Chr. 11 - 113687546 - 113709811 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |