Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1088 | LINC01088 | Anti-sense | Chr. 4 - 78971748 - 79308799 on NCBI Build GRCh38 | ||
Downstream |
N(alpha)-acetyltransferase 11, NatA catalytic subunit |
NAA11 |
ARD1B, ARD2, hARD2 |
17.6kb | Sense | Chr. 4 - 79155237 - 79326058 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |