Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
CTAGE family member 10, pseudogene |
CTAGE10P | -82.0kb | Anti-sense | Chr. 13 - 49890409 - 49893380 on NCBI Build GRCh38 | |
Downstream | deleted in lymphocytic leukemia 2 (non-protein coding) | DLEU2 |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
6.7kb | Sense | Chr. 13 - 49982552 - 50125541 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |