Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein FRG1-like | LOC102723780 | -967.2kb | Sense | Chr. 22 - 10940597 - 10961529 on NCBI Build GRCh38 | |
Downstream | olfactory receptor family 11 subfamily H member 1 | OR11H1 |
OR11H12, OR22-1 |
3.6mb | Anti-sense | Chr. 22 - 15528159 - 15529139 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |