Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GRB2 associated regulator of MAPK1 subtype 2 | GAREM2 |
FAM59B, GAREML |
Sense | Chr. 2 - 26173091 - 26202405 on NCBI Build GRCh38 | |
Downstream |
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit |
HADHB |
ECHB, MSTP029, MTPB, TP-BETA |
48.3kb | Sense | Chr. 2 - 26244748 - 26290465 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |