Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 15 | METTL15 | METT5D1 | Sense | Chr. 11 - 28108251 - 28527041 on NCBI Build GRCh38 | |
Downstream | microRNA 8068 | MIR8068 | hsa-mir-8068 | 351.6kb | Anti-sense | Chr. 11 - 28477481 - 28477548 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |