Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pleckstrin homology domain containing O2 | PLEKHO2 |
PLEKHQ1, PP1628, pp9099 |
-80.3kb | Anti-sense | Chr. 15 - 64841883 - 64868002 on NCBI Build GRCh38 |
Downstream |
spastic paraplegia 21 (autosomal recessive, Mast syndrome) |
SPG21 |
ACP33, BM-019, GL010, MAST |
13.0kb | Sense | Chr. 15 - 64963021 - 64989946 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |