Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100507291 | LOC100507291 | -24.4kb | Sense | Chr. 3 - 139389803 - 139583319 on NCBI Build GRCh38 | |
Downstream | calsyntenin 2 | CLSTN2 |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
325.2kb | Sense | Chr. 3 - 139935185 - 140577397 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |