Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |