Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100506023 | LOC100506023 | -10.0kb | Sense | Chr. 1 - 173235060 - 173477155 on NCBI Build GRCh38 | |
Downstream | solute carrier family 9 member C2 (putative) | SLC9C2 | SLC9A11 | 11.1kb | Sense | Chr. 1 - 173500465 - 173603094 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |