Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RNA, U2 small nuclear 1 |
RNU2-1 |
RNU2, U2 |
-123.5kb | Sense | Chr. 17 - 43233790 - 43233977 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 910 | LINC00910 | 9.3kb | Sense | Chr. 17 - 43369845 - 43388898 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |