Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK095500.1 | - | 1 | 2143 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | EF-hand calcium binding domain 2 | EFCAB2 |
CFAP200, DRC8 |
-581.9kb | Sense | Chr. 1 - 244969739 - 245125228 on NCBI Build GRCh38 |
Downstream | SET and MYND domain containing 3 | SMYD3 |
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1 |
39.9kb | Anti-sense | Chr. 1 - 245749340 - 246507342 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |