Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FGFR1OP N-terminal like | FOPNL |
C16orf63, FOR20, PHSECRG2 |
-178.3kb | Anti-sense | Chr. 16 - 15865719 - 15888649 on NCBI Build GRCh38 |
Downstream | ATP binding cassette subfamily C member 6 | ABCC6 |
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7 |
80.6kb | Anti-sense | Chr. 16 - 16149565 - 16223616 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |