Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 1233-1 | MIR1233-1 |
MIR1233, MIRN1233, hsa-mir-1233, hsa-mir-1233-1 |
-33.3kb | Sense | Chr. 15 - 34382069 - 34382150 on NCBI Build GRCh38 |
Downstream | golgin A8 family member B | GOLGA8B | GOLGA5 | 105.0kb | Sense | Chr. 15 - 34525276 - 34583651 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |