Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ring finger protein 5 pseudogene 1 | RNF5P1 | -237.5kb | Anti-sense | Chr. 8 - 38600173 - 38601257 on NCBI Build GRCh38 | |
Downstream | pleckstrin homology domain containing A2 | PLEKHA2 | TAPP2 | 61.2kb | Sense | Chr. 8 - 38901235 - 38973912 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |