Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PARK2 coregulated like | PACRGL | C4orf28 | -350.5kb | Anti-sense | Chr. 4 - 20696282 - 20757766 on NCBI Build GRCh38 |
Downstream | microRNA 7978 | MIR7978 | hsa-mir-7978 | 345.9kb | Sense | Chr. 4 - 21464700 - 21464758 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |