Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ELKS/RAB6-interacting/CAST family member 2 | ERC2 |
CAST, CAST1, ELKSL, SPBC110, Spc110 |
Sense | Chr. 3 - 55508311 - 56468473 on NCBI Build GRCh38 | |
Downstream | ERC2 intronic transcript 1 | ERC2-IT1 |
C1orf1, C3orf51, Po42 |
68.8kb | Sense | Chr. 3 - 55657206 - 55659469 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |