Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | galectin 12 | LGALS12 |
GAL12, GRIP1 |
-20.1kb | Sense | Chr. 11 - 63506052 - 63516774 on NCBI Build GRCh38 |
Downstream | HRAS like suppressor 2 | HRASLS2 | PLA1/2-2 | 12.8kb | Anti-sense | Chr. 11 - 63552759 - 63565072 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |