Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neurexophilin 3 | NXPH3 | NPH3 | -25.1kb | Sense | Chr. 17 - 49575936 - 49583810 on NCBI Build GRCh38 |
Downstream | solute carrier family 35 member B1 | SLC35B1 | UGTREL1 | 87.7kb | Anti-sense | Chr. 17 - 49700934 - 49708173 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |