Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 1273g | MIR1273G | -75.0kb | Sense | Chr. 1 - 52940314 - 52940413 on NCBI Build GRCh38 | |
Downstream | podocan | PODN |
PCAN, SLRR5A |
43.3kb | Sense | Chr. 1 - 53062052 - 53085502 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |