Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522125.2 | XP_011520427.1 | 1 | 493 | 59 | |
XM_017022687.1 | XP_016878176.1 | 1 | 493 | 59 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
proline, histidine and glycine rich 1 |
PHGR1 | Sense | Chr. 15 - 40351033 - 40356434 on NCBI Build GRCh38 | ||
Downstream | kinetochore localized astrin/SPAG5 binding protein | KNSTRN |
C15orf23, HSD11, SKAP, TRAF4AF1 |
27.5kb | Sense | Chr. 15 - 40382721 - 40394288 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |